Skip to Content
Merck
All Photos(1)

Documents

36156

Supelco

Methabenzthiazuron

PESTANAL®, analytical standard

Synonym(s):

1-(Benzothiazol-2-yl)-1,3-dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N3OS
CAS Number:
Molecular Weight:
221.28
Beilstein:
196633
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)N(C)c1nc2ccccc2s1

InChI

1S/C10H11N3OS/c1-11-9(14)13(2)10-12-7-5-3-4-6-8(7)15-10/h3-6H,1-2H3,(H,11,14)

InChI key

RRVIAQKBTUQODI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methabenzthiazuron is commonly used to control weeds and grasses, particularly in cereals, legumes and maize fields. It is regarded as very toxic for the environment and aquatic organisms. Mode of action of the herbicide involves inhibition of the ′Hill reaction′.

Application

Methabenzthiazuron may be used as an analytical reference standard for the determination of the analyte in drinking water, soil and fruits using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of the herbicide methabenzthiazuron on the synthesis of plastidic lipids in Hordeum vulgare seedlings.
Kleudgen HK
Pesticide Biochemistry and Physiology, 9(1), 57-60 (1978)
Determination of phenylurea herbicides in drinking water.
Hamada M and Wintersteiger R
J. Planar Chromatogr., 15(1), 11-18 (2002)
Methabenzthiazuron degradation with illuminated TiO2 aqueous suspensions. Kinetic and reactional pathway investigations.
Mezioud N, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 288, 13-22 (2014)
H P Malkomes
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Zweite naturwissenschaftliche Abteilung: Mikrobiologie der Landwirtschaft der Technologie und des Umweltschutzes, 134(7), 573-586 (1979-01-01)
In a field experiment the herbicides U 46 PD-Fluid (dichlorprop salt) and Tribunil (methabenzthiazuron) were applied in April 1976 post-emergence to winter wheat cultivated in 3 different soils. After 4 days, 1, 2 and 3 months samples were taken from
How to overcome matrix effects in the determination of pesticides in fruit by HPLC ESI MS MS.
Bester K, et al.
Fresenius Journal of Analytical Chemistry, 371(4), 550-555 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service