Skip to Content
Merck
All Photos(1)

Key Documents

36122

Supelco

Butoxycarboxim

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4S
CAS Number:
Molecular Weight:
222.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)O\N=C(/C)C(C)S(C)(=O)=O

InChI

1S/C7H14N2O4S/c1-5(6(2)14(4,11)12)9-13-7(10)8-3/h6H,1-4H3,(H,8,10)/b9-5+

InChI key

CTJBHIROCMPUKL-WEVVVXLNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butoxycarboxim belongs to N-methylcarbamate (NMC) class of pesticides, widely used in agriculture.[1]

Application

Butoxycarboxim may be used as an analytical reference standard for the determination of the analyte in soil, plant and air samples,[1] natural waters,[2] and agricultural products[3] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An enzyme-linked chemiluminescent immunoassay developed for detection of Butocarboxim from agricultural products based on monoclonal antibody.
Fang Q, et al.
Food Chemistry, 166(1), 372-379 (2015)
High-performance liquid chromatographic methods for the determination of N-methylcarbamate pesticides in water. soil, plants and air.
McGarvey BD
Journal of Chromatography A, 642(1-2), 89-105 (1993)
Simultaneous determination of acidic and non-acidic pesticides in natural waters by liquid chromatography-mass spectrometry.
Di Corcia A, et al.
Journal of Chromatography A, 878(1), 87-98 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service