Skip to Content
Merck
All Photos(1)

Key Documents

35826

Supelco

4-Chlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OH
CAS Number:
Molecular Weight:
128.56
Beilstein:
507004
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 49.8 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

220 °C (lit.)

mp

42-45 °C

density

1.306 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Oc1ccc(Cl)cc1

InChI

1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

WXNZTHHGJRFXKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chlorophenol may be used as an analytical reference standard for the determination of the analyte in environmental water, soil and in emissions of waste incineration plant by various chromatography methods.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Hygroskopic

Other Notes

Ion Mobility: TWCCSN2 value of 118.8 Å2 [M-H]-

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 35826 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Direct determination of chlorophenols in environmental water samples by hollow fiber supported ionic liquid membrane extraction coupled with high-performance liquid chromatography.
Peng JF, et al.
Journal of Chromatography A, 1139(2), 165-170 (2007)
Identification and quantification of volatile organic components in emissions of waste incineration plants.
Jay K and Stieglitz L
Chemosphere, 30(7), 1249-1260 (1995)
Determination of chlorophenols in soils using accelerated solvent extraction combined with solid-phase microextraction.
Wennrich L, et al.
Analytical Chemistry, 72(3), 546-551 (2000)
Qing Wen et al.
Journal of hazardous materials, 244-245, 743-749 (2012-11-28)
Bioelectrochemical systems (BESs), such as microbial fuel cells (MFCs) and microbial electrolysis cells (MECs), are generally regarded as a promising key technology to effectively treat wastewater and better exploit the renewable resource. This study explored the use of a BES
Marie Bank Nielsen et al.
Biodegradation, 24(1), 79-87 (2012-06-09)
A gram-positive bacterium Citricoccus nitrophenolicus (strain PNP1(T), DSM 23311(T), CCUG 59571(T)) isolated from a waste water treatment plant was capable of effectively degrading p-nitrophenol (pNP) as a source of carbon, nitrogen and energy for growth. Degradation of pNP required oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service