Skip to Content
Merck
All Photos(1)

Key Documents

35586

Supelco

PCB No 1

analytical standard

Synonym(s):

2-Chlorobiphenyl, 2-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9Cl
CAS Number:
Molecular Weight:
188.65
Beilstein:
1907934
Ballschmiter Number:
1
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccccc1-c2ccccc2

InChI

1S/C12H9Cl/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H

InChI key

LAXBNTIAOJWAOP-UHFFFAOYSA-N

General description

3-Chlorobiphenyl is a chlorinated biphenyl compound, which can be prepared, via phase transfer Gomberg-Bachmann-Hey reaction. It involves the condensation of 3-chlorobenzenediazonium tetrafluoroborate with 18-crown-6 in the presence of potassium acetate.[1]
Polychlorinated biphenyls (PCBs) are a class of highly toxic pollutants found in various environmental matrices and food products. They find a variety of applications as flame retardants, plasticizers, dielectric and heat transfer fluids, etc.[2]

Application

PCB No 1 may be used as an analytical reference standard for the quantification of the analyte in natural matrices using an on-line system for subcritical water extraction combined with high-performance liquid chromatography (HPLC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siham Beggah et al.
Microbial biotechnology, 1(1), 68-78 (2008-01-01)
Mutants were produced in the A-domain of HbpR, a protein belonging to the XylR family of σ(54)-dependent transcription activators, with the purpose of changing its effector recognition specificity from 2-hydroxybiphenyl (2-HBP, the cognate effector) to 2-chlorobiphenyl (2-CBP). Mutations were introduced
Hyeok Choi et al.
Environmental science & technology, 43(11), 4137-4142 (2009-07-03)
Reactive activated carbon (RAC) impregnated with palladized iron has been developed to effectively treat polychlorinated biphenyls (PCBs) in the environment by coupling adsorption and dechlorination of PCBs. In this study, we addressed the dechlorination reactivity and capacity of RAC toward
Christopher P O'Brien et al.
Environmental science & technology, 39(17), 6839-6844 (2005-09-30)
Incineration is commonly used to destroy polychlorinated biphenyl (PCB) wastes, but this method of treatment is not ideal for all mixed liquid wastes, especially those containing radioactive materials. Therefore, other remediation technologies are needed to efficiently treat these waste forms.
On-line coupling of subcritical water extraction with high-performance liquid chromatography via solid-phase trapping
Li B, et al.
Journal of Chromatography A, 873(2), 175-184 (2000)
O V Maltseva et al.
Biodegradation, 10(5), 363-371 (2000-06-28)
We studied the aerobic degradation of eight PCB congeners which comprise from 70 to 85% of the anaerobic dechlorination products from Aroclor 1242, including 2-, 4-, 2,4-, 2,6-, 2,2'-, 2,4'-, 2,2', 4-, and 2,4,4'-chlorobiphenyl (CB), and the biodegradation of their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service