Skip to Content
Merck

Skip To

35397

Bromchlorbuterol hydrochloride

VETRANAL®, analytical standard

Synonym(s):

1-(4-Amino-3-bromo-5-chlorophenyl)-2-tert-butylaminoethanol hydrochloride, Bromoclenbuterol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 159,00

About This Item

Empirical Formula (Hill Notation):
C12H18BrClN2O · HCl
CAS Number:
Molecular Weight:
358.10
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
6881138

€ 159,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC(C)(C)NCC(O)c1cc(Cl)c(N)c(Br)c1

InChI

1S/C12H18BrClN2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,10,16-17H,6,15H2,1-3H3;1H

InChI key

OWADKUMVZPHSHB-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
353999497232825
grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

format

neat

format

neat

format

neat

format

neat

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

application(s)

forensics and toxicology
pharmaceutical (small molecule)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®

product line

VETRANAL®


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters