Skip to Content
Merck
All Photos(2)

Key Documents

35389

Supelco

Triclabendazole sulfone

VETRANAL®, analytical standard

Synonym(s):

5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfonyl)-1H-benzimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€151.00

€151.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€151.00

About This Item

Empirical Formula (Hill Notation):
C14H9Cl3N2O3S
CAS Number:
Molecular Weight:
391.66
Beilstein:
18516857
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€151.00


Estimated to ship on21 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CS(=O)(=O)c1nc2cc(Cl)c(Oc3cccc(Cl)c3Cl)cc2[nH]1

InChI

1S/C14H9Cl3N2O3S/c1-23(20,21)14-18-9-5-8(16)12(6-10(9)19-14)22-11-4-2-3-7(15)13(11)17/h2-6H,1H3,(H,18,19)

InChI key

ZEIHWBIRYIXBSV-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Viviani et al.
Journal of veterinary pharmacology and therapeutics, 41(3), 476-484 (2018-02-22)
Parasitic diseases have a significant impact on livestock production. Nematodicidal drugs, such as fenbendazole (FBZ) or its oxidized metabolite oxfendazole (OFZ), can be used along with the trematodicidal triclabendazole (TCBZ), to broaden the spectrum of anthelmintic activity. However, co-exposure to
Yong Joo Lee et al.
PloS one, 8(5), e64337-e64337 (2013-05-15)
The Ras-adenylyl cyclase-protein kinase A nutrient-sensing pathway controls metabolism, proliferation and resistance to stress in Saccharomyces cerevisiae. The genetic disruption of this pathway increases resistance to a variety of stresses. We show here that the pharmacological inhibition of this pathway
Anton Kaufmann et al.
Journal of the American Society for Mass Spectrometry, 28(12), 2705-2715 (2017-09-14)
Targeted residue screening requires the use of reference substances in order to identify potential residues. This becomes a difficult issue when using multi-residue methods capable of analyzing several hundreds of analytes. Therefore, the capability of in silico fragmentation based on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service