Skip to Content
Merck
All Photos(2)

Key Documents

35373

Supelco

Hydramethylnon

PESTANAL®, analytical standard

Synonym(s):

1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one 2-(1,4,5,6-tetrahydro-5,5-dimethyl-2-pyrimidinyl)hydrazone, NSC 379665, Tetrahydro-5,5-dimethyl-2(1H)-pyrimidinone[3-[4-(trifluoromethyl)phenyl]-1-[2-[4-(trifluoromethyl)phenyl]ethenyl]-2-propylidene]hydrazone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€148.00

€148.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€148.00

About This Item

Empirical Formula (Hill Notation):
C25H24F6N4
CAS Number:
Molecular Weight:
494.48
Beilstein:
6015162
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€148.00


Estimated to ship on23 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)CN\C(NC1)=N\N=C(\C=C\c2ccc(cc2)C(F)(F)F)/C=C/c3ccc(cc3)C(F)(F)F

InChI

1S/C25H24F6N4/c1-23(2)15-32-22(33-16-23)35-34-21(13-7-17-3-9-19(10-4-17)24(26,27)28)14-8-18-5-11-20(12-6-18)25(29,30)31/h3-14H,15-16H2,1-2H3,(H2,32,33,35)/b13-7+,14-8+

InChI key

IQVNEKKDSLOHHK-FNCQTZNRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A G Appel
Journal of economic entomology, 85(4), 1176-1183 (1992-08-01)
Gel and paste bait formulations containing boric acid or hydramethylnon were evaluated in field and laboratory experiments with the German cockroach, Blattella germanica (L.). Bait formulations contained 14-62% water by weight and lost the most water after 1 d of
Changlu Wang et al.
Journal of economic entomology, 99(3), 879-885 (2006-07-04)
This study assessed the cost and effectiveness of a building-wide cockroach integrated pest management (IPM) program compared with bait alone treatment in public housing. In total, 12 buildings (66 apartments) were treated and monitored for cockroach infestations over 7 mo.
L M Smith et al.
Journal of economic entomology, 90(5), 1232-1242 (1997-12-31)
The insecticide components of a previously developed integrated pest management system for smokybrown cockroaches, Periplaneta fuliginosa (Serville), were tested individually and at reduced application rates in 4 separate trials over 2 yr. A 2-bait combination (pellet and gel baits) did
R Gaigher et al.
Ecological applications : a publication of the Ecological Society of America, 22(5), 1405-1412 (2012-08-23)
Effective management of invasive ants is an important priority for many conservation programs but can be difficult to achieve, especially within ecologically sensitive habitats. This study assesses the efficacy and nontarget risk of a precision ant baiting method aiming to
Pâmela Decio et al.
Micron (Oxford, England : 1993), 45, 22-31 (2012-11-15)
The leaf-cut ants are important agricultural pest, because they can cause intense defoliation in plants and destroy large areas cultivated. Although there are several works for the control of these insects by examining the toxicity of natural chemical compounds on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service