Skip to Content
Merck
All Photos(1)

Key Documents

34969

Supelco

Hexane, mixture of isomers

≥85% isomers basis (GC), for preparative liquid chromatography, suitable for HPLC

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
grade:
for preparative liquid chromatography
Assay:
≥85% isomers basis (GC)
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
68-70 °C (lit.)

Product Name

Hexane, mixture of isomers, for preparative liquid chromatography, ≥85% isomers basis (GC)

grade

for preparative liquid chromatography

vapor density

3 (vs air)

Assay

≥85% isomers basis (GC)

form

liquid

autoignition temp.

437 °F

expl. lim.

7.5 %

technique(s)

HPLC: suitable

impurities

≤0.0005% free alkali (as NH3)
≤0.0005% non-volatile matter
≤0.01% water (Karl Fischer)
≤4.9% n-hexane (GC)

transmittance

210 nm, ≥50%
250 nm, ≥98%

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

density

0.672 g/mL at 25 °C (lit.)

application(s)

food and beverages

SMILES string

C(CCC)(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Mohraz et al.
Langmuir : the ACS journal of surfaces and colloids, 21(12), 5298-5306 (2005-06-01)
The development of model materials and image processing methods to directly visualize and quantify colloidal rod assembly by means of confocal laser scanning microscopy (CLSM) is reported. Monodisperse fluorescent colloidal rods are prepared by the uniaxial extensional deformation of sterically
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service