Skip to Content
Merck
All Photos(1)

Key Documents

34520

Supelco

Fipronil sulfide

PESTANAL®, analytical standard

Synonym(s):

5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethylsulfanyl-1H-pyrazole-3-carbonitrile, 5-Amino-3-cyano-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethylthio)pyrazole

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€220.00

€220.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€220.00

About This Item

Empirical Formula (Hill Notation):
C12H4Cl2F6N4S
CAS Number:
Molecular Weight:
421.15
Beilstein:
5835917
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

€220.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

NC1=C(SC(F)(F)F)C(C#N)=NN1C2=C(Cl)C=C(C(F)(F)F)C=C2Cl

InChI

1S/C12H4Cl2F6N4S/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)25-12(18,19)20/h1-2H,22H2

InChI key

FQXWEKADCSXYOC-UHFFFAOYSA-N

General description

Fipronil sulfide is a degradation product of the broad spectrum insecticide fipronil, which is found to be toxic to a variety of non-target organisms.[1]

Application

Fipronil sulfide may be used as a reference standard in the determination of fipronil sulfide in honey and pollen samples using off line solid phase extraction liquid chromatography-tandem mass spectrometry method.[2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Validation of an off line solid phase extraction liquid chromatography?tandem mass spectrometry method for the determination of systemic insecticide residues in honey and pollen samples collected in apiaries from NW Spain.
Garcia-Chao M, et al.
Analytica Chimica Acta, 672(1-2), 107-113 (2010)
Influence of fipronil compounds and rice-cultivation land-use intensity on macroinvertebrate communities in streams of southwestern Louisiana, USA.
Mize VS, et al.
Environmental Pollution (Barking, Essex : 1987), 152(2), 491-503 (2008)
Janet L Miller et al.
Science advances, 6(43) (2020-10-25)
Insecticides in streams are increasingly a global concern, yet information on safe concentrations for aquatic ecosystems is sparse. In a 30-day mesocosm experiment exposing native benthic aquatic invertebrates to the common insecticide fipronil and four degradates, fipronil compounds caused altered

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service