Skip to Content
Merck
All Photos(2)

Key Documents

34189

Supelco

Dimethyl phthalate-3,4,5,6-d4

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€222.00

€222.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€222.00

About This Item

Linear Formula:
C6D4(CO2CH3)2
CAS Number:
Molecular Weight:
198.21
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€222.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.515 (lit.)

bp

282 °C (lit.)

mp

2 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OC)c(c1[2H])C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i3D,4D,5D,6D

InChI key

NIQCNGHVCWTJSM-LNFUJOGGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service