Skip to Content
Merck
All Photos(2)

Key Documents

34184

Supelco

Sudan I-d5

analytical standard

Synonym(s):

1-Phenylazo-2-naphthol-d5

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€206.00

€206.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€206.00

About This Item

Linear Formula:
C6D5N=NC10H6OH
CAS Number:
Molecular Weight:
253.31
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€206.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[2H]c1c([2H])c([2H])c(\N=N\c2c(O)ccc3ccccc23)c([2H])c1[2H]

InChI

1S/C16H12N2O/c19-15-11-10-12-6-4-5-9-14(12)16(15)18-17-13-7-2-1-3-8-13/h1-11,19H/b18-17+/i1D,2D,3D,7D,8D

InChI key

MRQIXHXHHPWVIL-BWDNVODCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sudan I-d5 may be used as an internal standard for the determination of the azo dyes in feed by ultra-high performance liquid chromatography-tandem mass spectrometry.[1]

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>554.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 290 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of ten illegal azo dyes in feed by ultra-high performance liquid chromatography tandem mass spectrometry.
Piatkowska M, et al.
Journal of Veterinary Research, 61(3), 299-305 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service