Skip to Content
Merck
All Photos(1)

Key Documents

33977

Supelco

Foramsulfuron

PESTANAL®, analytical standard

Synonym(s):

2-[3-(4,6-Dimethoxy-2-pyrimidinyl)ureidosulfonyl]-4-(formamido)-N,N-dimethylbenzamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€105.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€105.00

About This Item

Empirical Formula (Hill Notation):
C17H20N6O7S
CAS Number:
Molecular Weight:
452.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€105.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[H]C(=O)Nc1ccc(C(=O)N(C)C)c(c1)S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

InChI

1S/C17H20N6O7S/c1-23(2)15(25)11-6-5-10(18-9-24)7-12(11)31(27,28)22-17(26)21-16-19-13(29-3)8-14(20-16)30-4/h5-9H,1-4H3,(H,18,24)(H2,19,20,21,22,26)

InChI key

PXDNXJSDGQBLKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Foramsulfuron is a sulfonylurea herbicide used in controlling a wide spectrum of weeds. Its mode of action involves inhibiting the enzyme activity of acetolactate synthase (ALS) which is involved in biosynthesis of branched amino acids.[1]

Application

Foramsulfuron may be used as a reference standard for the determination of foramsulfuron in water samples by solid-phase extraction and liquid chromatography-tandem mass spectrometry (SPE-LC-MS/MS).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photooxidation of foramsulfuron: Effects of char substances.
Vittoria PM, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 326, 16-20 (2016)
James T Brosnan et al.
Planta, 243(1), 149-159 (2015-09-12)
This is a first report of an Ala-205-Phe substitution in acetolactate synthase conferring resistance to imidazolinone, sulfonylurea, triazolopyrimidines, sulfonylamino-carbonyl-triazolinones, and pyrimidinyl (thio) benzoate herbicides. Resistance to acetolactate synthase (ALS) and photosystem II inhibiting herbicides was confirmed in a population of
Trace analysis of sulfonylurea herbicides in water samples by solid-phase extraction and liquid chromatography-tandem mass spectrometry.
Fenoll J
Talanta, 101, 273-282 (2012)
James T Brosnan et al.
Pest management science, 76(6), 2049-2057 (2020-01-17)
Indaziflam is an alkylazine herbicide used to control annual bluegrass (Poa annua L.). Several locations in the southeastern USA reported poor annual bluegrass control following treatment with indaziflam during autumn 2015. A series of controlled environment experiments were conducted to
Amit Paporisch et al.
Pesticide biochemistry and physiology, 138, 22-28 (2017-05-01)
Three sweet corn genotypes, two inbred lines (IBER001 and IBER002) and their hybrid (ER00X), differ in their phenotypic responses to several P450-metabolized herbicides, used in sweet corn, namely, foramsulfuron, iodosulfuron, rimsulfuron and tembotrione. Foramsulfuron is a sulfonylurea herbicide commonly formulated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service