Skip to Content
Merck
All Photos(2)

Key Documents

33888

Supelco

Ethofumesate-2-keto

PESTANAL®, analytical standard

Synonym(s):

3,3-Dimethyl-2-oxo-2,3-dihydrobenzofurane-5-yl methane sulfonate, Methane sulfonic acid 3,3-dimethyl-2-oxo-2,3-dihydrobenzofuran-5-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O5S
CAS Number:
Molecular Weight:
256.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(=O)Oc2ccc(OS(C)(=O)=O)cc12

InChI

1S/C11H12O5S/c1-11(2)8-6-7(16-17(3,13)14)4-5-9(8)15-10(11)12/h4-6H,1-3H3

InChI key

CXWYCAYNZXSHTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethofumesate-2-keto is a metabolite of the herbicide, ethofumesate used for the control of weeds in strawberry and beet crops. It inhibits the growth of meristems, retards cellular division and limits the formation of cuticules.

Application

Ethofumesate-2-keto may be used as an analytical reference standard for the determination of the analyte in strawberries and soil samples using quick, easy, cheap, effective, rugged, and safe (QuEChERS) method of extraction followed by gas chromatography (GC)/liquid chromatography (LC) coupled to tandem mass spectrometry (MS/MS) and two-dimensional gas chromatography time-of-flight mass spectrometry (GC × GC-TOF-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of pesticide residues in strawberries and soils by GC-MS/MS, LC-MS/MS and two-dimensional GC-time-of-flight MS comparing organic and integrated pest management farming.
Fernandes CV, et al.
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment, 31(2), 262-270 (2014)
Physiological basis for bermudagrass control with ethofumesate in seashore paspalum and St. Augustinegrass.
McCullough EP, et al.
Crop Science, 56(3), 1306-1313 (2016)
Degradation of ethofumesate in soil under laboratory conditions.
Kucharski M and Sadowski J
Polish Journal of Environmental Studies, 18(2), 1306-1313 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service