Skip to Content
Merck
All Photos(1)

Key Documents

33874

Supelco

Cyazofamid

PESTANAL®, analytical standard

Synonym(s):

4-Chloro-1-(dimethylaminosulfonyl)-5-(p-tolyl)imidazole-2-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€83.60

€83.60


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€83.60

About This Item

Empirical Formula (Hill Notation):
C13H13ClN4O2S
CAS Number:
Molecular Weight:
324.79
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€83.60


Estimated to ship on16 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CN(C)S(=O)(=O)n1c(nc(Cl)c1-c2ccc(C)cc2)C#N

InChI

1S/C13H13ClN4O2S/c1-9-4-6-10(7-5-9)12-13(14)16-11(8-15)18(12)21(19,20)17(2)3/h4-7H,1-3H3

InChI key

YXKMMRDKEKCERS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyazofamid is a fungicide exhibiting specific activity against diseases caused by oomycetes fungi and Plasmodiophora brassicae Woronin.[1]

Application

Cyazofamid can be used as an analytical reference standard for the determination of the analyte in grapes and wine samples[2][3] using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of nine fungicides in grape and wine using QuEChERS extraction and LC/MS/MS analysis.
Matinez G, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 98(1745-1751), 6-6 (2015)
Effects of cyazofamid against Plasmodiophora brassicae Woronin on Chinese cabbage.
Mitani S, et al.
Pest Management Science, 59(3), 287-293 (2003)
Im-Sook Song et al.
Journal of biochemical and molecular toxicology, 34(4), e22459-e22459 (2020-02-01)
We aimed to investigate the intestinal permeability and interaction of cyazofamid with clinically important transporters. The intestinal permeability of cyazofamid was low (0.21 ± 0.02 cm/s), and it is a substrate for P-glycoprotein (P-gp) with a Km value of 83.1 μM, indicated that P-gp
Multiresidue determination of 11 new fungicides in grapes and wines by liquid?liquid extraction/clean-up and programmable temperature vaporization injection with analyte protectants/gas chromatography/ion trap mass spectrometry.
Gonzalez-Rodriguez R M, et al.
Journal of Chromatography A, 1216(32), 6033-6042 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service