Skip to Content
Merck
All Photos(1)

Documents

33872

Supelco

Fenoxanil

PESTANAL®, analytical standard

Synonym(s):

N-(1-Cyano-1,2-dimethylpropyl)-2-(2,4-dichlorophenoxy)propionamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18Cl2N2O2
CAS Number:
Molecular Weight:
329.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)C(C)(NC(=O)C(C)Oc1ccc(Cl)cc1Cl)C#N

InChI

1S/C15H18Cl2N2O2/c1-9(2)15(4,8-18)19-14(20)10(3)21-13-6-5-11(16)7-12(13)17/h5-7,9-10H,1-4H3,(H,19,20)

InChI key

IUOKJNROJISWRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Fenoxanil is a systemic fungicide used for the control of rice blast.

Application

Fenoxanil may be used as an analytical reference standard for the determination of the analyte in:
  • Agricultural products using gas chromatography coupled to mass spectrometry (GC-MS).
  • Lemons, beans, and other vegetables using gas chromatography coupled to tandem mass spectrometry (GC-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 105 pesticide residues by GC/MS/MS.
Kim SW, et al.
Analytical Science and Technology, 23(4), 395-404 (2010)
Feng Zhu et al.
International journal of molecular sciences, 19(10) (2018-09-23)
Mesoporous silica nanoparticles (MSNs) can be used as carriers to deliver pesticides into plants, which is considered to be one method of improving the efficacy of pesticide usage in agricultural production. In the present work, MSNs with an average diameter
Marc Pihet et al.
BMC microbiology, 9, 177-177 (2009-08-26)
Aspergillus fumigatus is the most common agent of invasive aspergillosis, a feared complication in severely immunocompromised patients. Despite the recent commercialisation of new antifungal drugs, the prognosis for this infection remains uncertain. Thus, there is a real need to discover
Leesun Kim et al.
Molecules (Basel, Switzerland), 25(15) (2020-08-06)
An effective analytical method was optimized for residues including chlorpyrifos-methyl, deltamethrin, fenoxanil, thiobencarb and fludioxonil in mealworms, the larval form of Tenebrio molitor. They are listed for pest control during wheat cultivation and can be found in wheat-bran feed for
A New Systemic Fungicide for the Control of Rice Blast: Fenoxanil [J].
Lin L, et al.
Agrochemicals, 7(4), 13-13 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service