Skip to Content
Merck
All Photos(1)

Documents

33759

Supelco

Microcystin-LY solution

2-8 μg/mL in methanol, analytical standard

Synonym(s):

Algae bloom toxin, Biotoxin, Cyanobacterial toxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H71N7O13
CAS Number:
Molecular Weight:
1002.16
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

2-8 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@@H](Cc1ccccc1)[C@@H](C)\C=C(C)\C=C\[C@@H]2NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](C)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](C)NC(=O)C(=C)N(C)C(=O)CC[C@@H](NC(=O)[C@H]2C)C(O)=O)C(O)=O

InChI

1S/C52H71N7O13/c1-28(2)24-40-50(67)58-44(52(70)71)32(6)46(63)56-41(26-36-17-19-37(60)20-18-36)49(66)54-38(21-16-29(3)25-30(4)42(72-10)27-35-14-12-11-13-15-35)31(5)45(62)55-39(51(68)69)22-23-43(61)59(9)34(8)48(65)53-33(7)47(64)57-40/h11-21,25,28,30-33,38-42,44,60H,8,22-24,26-27H2,1-7,9-10H3,(H,53,65)(H,54,66)(H,55,62)(H,56,63)(H,57,64)(H,58,67)(H,68,69)(H,70,71)/b21-16+,29-25+/t30-,31-,32-,33+,38-,39+,40-,41-,42-,44+/m0/s1

InChI key

SIGQAYSWORHPPH-WSTSHCHISA-N

General description

Microcystin-LY is generally produced by cyanobacteria in freshwater systems and is found as a toxic analog of microcystin.

Application

Microcystin-LY may be used as an analytical reference standard for the determination of the analyte in raw and treated waters using high-performance liquid chromatography with photo-diode array detection (HPLC-PDA).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Rudolph-Böhner et al.
Biological chemistry Hoppe-Seyler, 374(8), 635-640 (1993-08-01)
Two toxins, a main component A and a minor component B, were isolated from the freshwater cyanobacterium Microcystis aeruginosa (strain 298) and characterized in their chemical structure by amino-acid analysis, configurational analysis, by FAB-MS and 1H-NMR spectroscopy. The acid hydrolysate
Fatima El Khalloufi et al.
Ecotoxicology and environmental safety, 79, 199-205 (2012-01-31)
The cyanobacterial toxins microcystins (MC) are known to affect many processes in plants. Their presence in the water used for irrigation may have considerable impact on the survivorship, growth and development of plants. In this study, a crude extract of
Hiroshi Ishii et al.
Water research, 38(11), 2667-2676 (2004-06-23)
A bacterium termed 7CY, capable of decomposing cyanobacterial toxins, was isolated from surface water sample of Lake Suwa and degradation of microcystin-RR and nodularin-Har was investigated. The isolated 7CY was a gram-negative, aerobic bacillus, and a member of a genus
S Rudolph-Böhner et al.
FEBS letters, 349(3), 319-323 (1994-08-08)
The three-dimensional structure of the two hepatotoxic microcystins LR and LY has been determined by two-dimensional nuclear magnetic resonance (2D NMR) spectroscopy and distance geometry calculations. For the microcystin LY a single family of highly convergent structures was obtained. This
C J Ward et al.
Journal of applied microbiology, 86(5), 874-882 (1999-05-29)
Microcystins (MC) are a group of over 60 cyclic heptapeptide hepatotoxins produced by cyanobacteria. The 1-octanol/water partition coefficients (log P) of MC-LR, -LY, -LW and -LF have been estimated by HPLC to be 2.16, 2.92, 3.46 and 3.56, respectively. Their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service