Skip to Content
Merck
All Photos(2)

Key Documents

33652

Supelco

2-Isopropyl-d7-thioxanthen-9-one

analytical standard

Synonym(s):

ITX-d7

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€441.00

€441.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€441.00

About This Item

Empirical Formula (Hill Notation):
C16D7H7OS
CAS Number:
Molecular Weight:
261.39
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€441.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

mass shift

M+7

SMILES string

[2H]C([2H])([2H])C([2H])(c1ccc2Sc3ccccc3C(=O)c2c1)C([2H])([2H])[2H]

InChI

1S/C16H14OS/c1-10(2)11-7-8-15-13(9-11)16(17)12-5-3-4-6-14(12)18-15/h3-10H,1-2H3/i1D3,2D3,10D

InChI key

KTALPKYXQZGAEG-SVMCCORHSA-N

Application

2-Isopropyl-d7-thioxanthen-9-one may be used as an internal standard for the analysis of UV ink photoinitiators in packaged food using QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) method followed by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of UV ink photoinitiators in packaged food by fast liquid chromatography at sub-ambient temperature coupled to tandem mass spectrometry.
Gallart-Ayala H, et al.
Journal of Chromatography A, 1218(3), 459-466 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service