Skip to Content
Merck
All Photos(3)

Documents

335940

Sigma-Aldrich

Xylene Cyanol FF

Dye content ≥75 %

Synonym(s):

Acid blue 147, Cyanol FF, XC, Xylene Cyanole FF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27N2NaO6S2
CAS Number:
Molecular Weight:
538.61
Colour Index Number:
42135
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

≥96.0% (HPLC)

Quality Level

form

powder or crystals

composition

Dye content, ≥75%

mp

295 °C (dec.) (lit.)

solubility

water: 1 mg/mL, blue

λmax

615 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCNc1ccc(cc1C)C(=C2/C=C\C(=N/CC)C(C)=C2)\c3ccc(cc3S(O)(=O)=O)S([O-])(=O)=O

InChI

1S/C25H28N2O6S2.Na/c1-5-26-22-11-7-18(13-16(22)3)25(19-8-12-23(27-6-2)17(4)14-19)21-10-9-20(34(28,29)30)15-24(21)35(31,32)33;/h7-15,26H,5-6H2,1-4H3,(H,28,29,30)(H,31,32,33);/q;+1/p-1/b25-19-,27-23+;

InChI key

VVLFAAMTGMGYBS-AHRQKMASSA-M

Related Categories

General description

Xylene cyanol is often used as a tracking dye during agarose or polyacrylamide gel electrophoresis. It has a slightly negative charge and will migrate in the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The tracking dye typically migrates with the DNA molecules around 5kb. Xylene cyanol may also be used in loading buffer that facilitates the observation of the sample during gel loading and electrophoresis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discriminatory power of agarose gel electrophoresis in DNA fragments analysis.
Lee SV and Abdul RB et al.
Gel Electrophoresis of Nucleic Acids: A Practical Approach (2012)
Kumar A and Garg N
Genetic Engineering (2005)
Frequency of the BRCA1 185delAG mutation among Jewish women with ovarian cancer and matched population controls.
Muto MG
Cancer Research, 56(6), 1250-1252 (1996)
Katrina M Morris et al.
BMC genomics, 16, 1017-1017 (2015-11-28)
The Tasmanian devil (Sarcophilus harrisii) is being threatened with extinction in the wild by a disease known as devil facial tumour disease (DFTD). In order to prevent the spread of this disease a thorough understanding of the Tasmanian devil immune
Luyan Zhang et al.
G3 (Bethesda, Md.), 5(3), 427-439 (2015-01-17)
In this study, we considered four categories of molecular markers based on the number of distinguishable alleles at the marker locus and the number of distinguishable genotypes in clonal F1 progenies. For two marker loci, there are nine scenarios that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service