Skip to Content
Merck
All Photos(1)

Documents

32872

Supelco

Alanycarb

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N3O4S2
CAS Number:
Molecular Weight:
399.53
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)CCN(Cc1ccccc1)SN(C)C(=O)O\N=C(\C)SC

InChI

1S/C17H25N3O4S2/c1-5-23-16(21)11-12-20(13-15-9-7-6-8-10-15)26-19(3)17(22)24-18-14(2)25-4/h6-10H,5,11-13H2,1-4H3/b18-14-

InChI key

GMAUQNJOSOMMHI-JXAWBTAJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alanycarb is classified under the oxime carbamate group of insecticides. Its mode of action involves the inhibition of cholinesterase enzyme.

Application

Alanycarb may be used as an analytical reference standard for the determination of the analyte in:
  • Olive oil samples by fast liquid chromatography-electrospray time-of-flight mass spectrometry (LC-TOFMS).
  • Vegetables and fruit juices by LC coupled to tandem mass spectrometry (MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic Pathways of Agrochemicals: Insecticides and fungicides, 1 (1998)
Evaluation of two sample treatment methodologies for large-scale pesticide residue analysis in olive oil by fast liquid chromatography-electrospray mass spectrometry.
Gilbert-Lopez B, et al.
Journal of Chromatography A, 1217(24), 3736-3747 (2010)
Finding of pesticides in fashionable fruit juices by LC-MS/MS and GC-MS/MS.
Tran K, et al.
Food Chemistry, 134(4), 2398-2405 (2012)
Validation and uncertainty study of a comprehensive list of 160 pesticide residues in multi-class vegetables by liquid chromatography-tandem mass spectrometry.
Kmellar B, et al.
Journal of Chromatography A, 1215(1-2), 37-50 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service