Skip to Content
Merck
All Photos(2)

Key Documents

32584

Supelco

Tembotrione metabolite AE 1417268

PESTANAL®, analytical standard

Synonym(s):

2-{2-Chloro-4-mesyl-3-[(2,2,2-trifluoroethoxy)methyl]benzoyl}-4,6-dihydroxycyclohexane-1,3-dione

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€189.00

€189.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€189.00

About This Item

Empirical Formula (Hill Notation):
C17H16ClF3O8S
CAS Number:
Molecular Weight:
472.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€189.00


Estimated to ship on17 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CS(=O)(=O)c1ccc(c(Cl)c1COCC(F)(F)F)C(=O)C2C(=O)C(O)CC(O)C2=O

InChI

1S/C17H16ClF3O8S/c1-30(27,28)11-3-2-7(13(18)8(11)5-29-6-17(19,20)21)14(24)12-15(25)9(22)4-10(23)16(12)26/h2-3,9-10,12,22-23H,4-6H2,1H3

InChI key

KGPIRVWYAUCFAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service