Skip to Content
Merck
All Photos(2)

Key Documents

32527

Supelco

Acequinocyl

PESTANAL®, analytical standard

Synonym(s):

2-(Acetyloxy)-3-dodecyl-1,4-naphthalenedione, 3-Acetoxy-2-dodecyl-1,4-naphthoquinone, 3-Dodecyl-1,4-dihydro-1,4-dioxo-2-naphthyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€125.00

€125.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€125.00

About This Item

Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Beilstein:
2016033
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€125.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCC1=C(OC(C)=O)C(=O)c2ccccc2C1=O

InChI

1S/C24H32O4/c1-3-4-5-6-7-8-9-10-11-12-17-21-22(26)19-15-13-14-16-20(19)23(27)24(21)28-18(2)25/h13-16H,3-12,17H2,1-2H3

InChI key

QDRXWCAVUNHOGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

AKD-2023, 3-dodecyl-1,4-dihydro-1,4-dioxo-2-naphthyl acetate commonly known as acequinocyl, is a novel miticide . Its miticidal activity is very efficient against economically important spider mites. Its mode of action is via inhibition of mitochondrial electron transfer at complex III.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 - STOT SE 1 Inhalation

Target Organs

Blood, Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AKD-2023: A novel miticide. Biological activity and mode of action.
Kinoshita, Sachiko, et al.
Pest Management Science, 55 (6), 659-660 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service