Skip to Content
Merck
All Photos(1)

Key Documents

32524

Supelco

Isotianil

PESTANAL®, analytical standard

Synonym(s):

3,4-Dichloro-2′-cyano-1,2-thiazole-5-carboxanilide, 3,4-Dichloro-N-(2-cyanophenyl)-5-isothiazolecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H5Cl2N3OS
CAS Number:
Molecular Weight:
298.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1nsc(C(=O)Nc2ccccc2C#N)c1Cl

InChI

1S/C11H5Cl2N3OS/c12-8-9(18-16-10(8)13)11(17)15-7-4-2-1-3-6(7)5-14/h1-4H,(H,15,17)

InChI key

WLPCAERCXQSYLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isotianil is a commonly used pesticide which activates defence responses in plants against a wide range of pathogens.[1][2][3] It is used in preventing the disease named rice blast, caused by rice blast fungi.[3]

Application

Isotianil has been used as a reference standard for the determination of isotianil in brown rice and soil using supercritical fluid extraction and gas chromatography/mass spectrometry method.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of analytical method for Isotianil in agricultural commodities using GC-NPD and GC-MSD.
Do A-J, et al.
Journal of Chemical and Pharmaceutical Sciences , 25(6), 364-369 (2012)
Determination of isotianil in brown rice and soil using supercritical fluid extraction and gas chromatography/mass spectrometry.
Yoshida T, et al.
Analytical Sciences, 29(9), 919-922 (2013)
Applied development of a novel fungicide isotianil (Stout).
Ogawa M, et al.
Journal of Chemical and Pharmaceutical Sciences , 1, 1-15 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service