Skip to Content
Merck
All Photos(2)

Documents

32518

Supelco

Sulfadiazine-(phenyl-13C6)

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(2-pyrimidinyl)benzene-13C6-sulfonamide, N1-(Pyrimidin-2-yl)sulfanilamide13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C4H10N4O2S
CAS Number:
Molecular Weight:
256.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+6

SMILES string

N[13c]1[13cH][13cH][13c]([13cH][13cH]1)S(=O)(=O)Nc2ncccn2

InChI

1S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7H,11H2,(H,12,13,14)/i2+1,3+1,4+1,5+1,8+1,9+1

InChI key

SEEPANYCNGTZFQ-AHBHZWPESA-N

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ming Xue et al.
Analytical and bioanalytical chemistry, 412(1), 203-222 (2019-12-06)
This study examines an improved and simplified method for solid-phase extraction (SPE), which offers rapid and accurate determination and identification of 44 pharmaceutically active compounds using ultra-performance liquid chromatography (UPLC) and tandem mass spectrometry (MS/MS). The common active compounds include

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service