Skip to Content
Merck
All Photos(2)

Key Documents

32496

Supelco

1-Hydroxyibuprofen

VETRANAL®, analytical standard

Synonym(s):

2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€301.00

€301.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€301.00

About This Item

Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
Beilstein:
22307543
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€301.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)C(O)c1ccc(cc1)C(C)C(O)=O

InChI

1S/C13H18O3/c1-8(2)12(14)11-6-4-10(5-7-11)9(3)13(15)16/h4-9,12,14H,1-3H3,(H,15,16)

InChI key

RMOQYHYFRKTDRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hydroxyibuprofen is one of the metabolites of ibuprofen, which is classified under the non-steroidal anti-inflammatory drugs (NSAIDs) group of compounds.[1]

Application

1-Hydroxyibuprofen has been used as an analytical reference standard for the determination of the analyte in:
  • Sewage sludge samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction with on-line solid-phase extract purification and pre-concentration, followed by analysis using liquid chromatography coupled to electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS) operating in multiple reaction monitoring (MRM) mode of detection.[1]


It may be used as an analytical reference standard for the determination of the analyte in:
  • Human urine samples by ultrasound-assisted emulsification microextraction (USAEME) and ultra-high performance liquid chromatography (UHPLC) with (ESI-MS/MS) with (MRM) detection.[2]
  • Wastewater samples by hollow fiber liquid phase microextraction (HF-LPME) followed by LC-ESI-MS/MS.[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound-assisted emulsification microextraction combined with ultra-high performance liquid chromatography-tandem mass spectrometry for the analysis of ibuprofen and its metabolites in human urine.
Magiera S and Gulmez S
Journal of Pharmaceutical and Biomedical Analysis, 92, 193-202 (2014)
Behaviour of nonsteroidal anti-inflammatory drugs and eight of their metabolites during wastewater treatment studied by hollow fibre liquid phase microextraction and liquid chromatography mass spectrometry.
Larsson E, et al.
The Science of the Total Environment, 485, 300-308 (2014)
Innovative combination of QuEChERS extraction with on-line solid-phase extract purification and pre-concentration, followed by liquid chromatography-tandem mass spectrometry for the determination of non-steroidal anti-inflammatory drugs and their metabolites in sewage sludge.
Rossini D, et al.
Analytica Chimica Acta, 935, 269-281 (2016)
Aleksandar Lolić et al.
The Science of the total environment, 508, 240-250 (2014-12-08)
The occurrence of seven pharmaceuticals and two metabolites belonging to non-steroidal anti-inflammatory drugs and analgesics therapeutic classes was studied in seawaters. A total of 101 samples covering fourteen beaches and five cities were evaluated in order to assess the spatial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service