Skip to Content
Merck
All Photos(2)

Key Documents

32429

Supelco

Prothioconazole-desthio

PESTANAL®, analytical standard

Synonym(s):

α-(1-Chlorocyclopropyl)-α-[(2-chlorophenyl)methyl]-1H-1,2,4-triazole-1-ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
€278.00

€278.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
20 MG
€278.00

About This Item

Empirical Formula (Hill Notation):
C14H15Cl2N3O
CAS Number:
Molecular Weight:
312.19
Beilstein:
13200866
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€278.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(Cc1ccccc1Cl)(Cn2cncn2)C3(Cl)CC3

InChI

1S/C14H15Cl2N3O/c15-12-4-2-1-3-11(12)7-14(20,13(16)5-6-13)8-19-10-17-9-18-19/h1-4,9-10,20H,5-8H2

InChI key

HHUQPWODPBDTLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prothioconazole-desthio is a major chiral metabolite of prothioconazole fungicide.[1]

Application

Prothioconazole-desthio may be used as an analytical reference standard for the determination of the analyte in:
  • Rapeseed oil samples by gas chromatography coupled to electron capture detection (GC-ECD).[2]
  • Food samples of animal origin by high performance liquid chromatography combined with electrospray ionization-tandem mass spectrometry (HPLC-ESI-MS/MS).[3]
  • Honeybees by dispersive solid-phase extraction (dSPE) combined with LC coupled to electron ionization (EI) MS/MS operating in multiple reaction monitoring (MRM) mode of detection.[4]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry?Honeybee poisoning incidents.
Kiljanek T, et al.
Journal of Chromatography A, 1435(11), 100-114 (2016)
An improved extraction method of rapeseed oil sample preparation for the subsequent determination in it of azole class fungicides by gas chromatography.
Zayats MF, et al.
Analytical Chemistry Research, 3(37), 37-45 (2015)
Simultaneous enantioselective determination of the chiral fungicide Prothioconazole and its major chiral metabolite Prothioconazole-desthio in food and environmental samples by Ultraperformance liquid chromatography-tandem mass spectrometry.
Zhang Z, et al.
Journal of Agricultural and Food Chemistry, 65(37), 8241-8247 (2017)
Approach for pesticide residue analysis for metabolite prothioconazole-desthio in animal origin food.
Liu H, et al.
Journal of Agricultural and Food Chemistry, 65(11), 2481-2487 (2017)
Iza Matarashvili et al.
Journal of separation science, 38(24), 4173-4179 (2015-10-27)
The separation of the stereoisomers of 23 chiral basic agrochemicals was studied on six different polysaccharide-based chiral columns in high-performance liquid chromatography with various polar organic mobile phases. Along with the successful separation of analyte stereoisomers, emphasis was placed on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service