Skip to Content
Merck

Skip To

32405

Flupyrsulfuron-methyl sodium

PESTANAL®, analytical standard

Synonym(s):

Methyl 2-[(4,6-dimethoxypyrimidin-2-ylcarbamoyl)sulfamoyl]-6-(trifluoromethyl)nicotinate monosodium salt monosodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 478,00

About This Item

Empirical Formula (Hill Notation):
C15H13F3N5NaO7S
CAS Number:
Molecular Weight:
487.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€ 478,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=C([N-]S(=O)(C1=NC(C(F)(F)F)=CC=C1C(OC)=O)=O)NC2=NC(OC)=CC(OC)=N2.[Na+]

InChI

1S/C15H14F3N5O7S.Na/c1-28-9-6-10(29-2)21-13(20-9)22-14(25)23-31(26,27)11-7(12(24)30-3)4-5-8(19-11)15(16,17)18;/h4-6H,1-3H3,(H2,20,21,22,23,25);/q;+1/p-1

InChI key

JKPSVOHVUGMYGH-UHFFFAOYSA-M

General description

Flupyrsulfuron-methyl sodium is a post-emergence herbicide, which is used in controlling annual broadleaf weeds in cereal plantation.

Application

Flupyrsulfuron-methyl sodium may be used as a reference standard in the determination of flupyrsulfuron-methyl sodium in soil samples using high performance liquid chromatography coupled with ultraviolet detector (HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
329183791633197
format

neat

format

neat

format

neat

format

neat

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

product line

PESTANAL®

product line

PESTANAL®

product line

PESTANAL®

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


pictograms

Environment

signalword

Warning

hcodes

pcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters