Skip to Content
Merck
All Photos(1)

Key Documents

32388

Supelco

(±)-Metoprolol-(isopropyl-d7) (+)-tartrate salt

VETRANAL®, analytical standard

Synonym(s):

(±)-Metoprolol-(isopropyl-d7) hemi-(+)-tartrate salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€719.00

€719.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€719.00

About This Item

Empirical Formula (Hill Notation):
C30D14H36N2O6 · C4H6O6
CAS Number:
Molecular Weight:
698.90
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€719.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical
veterinary

format

neat

storage temp.

2-8°C

SMILES string

OC(CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])COC1=CC=C(CCOC)C=C1.OC([C@H](O)[C@@H](O)C(O)=O)=O.OC(CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])COC2=CC=C(CCOC)C=C2

InChI

1S/2C15H25NO3.C4H6O6/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-1(3(7)8)2(6)4(9)10/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1/i2*1D3,2D3,12D;

InChI key

YGULWPYYGQCFMP-UGWAQTNBSA-N

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magda Caban et al.
Analytical methods : advancing methods and applications, 12(30), 3854-3864 (2020-07-24)
The study aimed to show the limitations and advantages of the use of stable isotope labeled internal standards (SILISs) for the quantification of small polar compounds (ibuprofen, diclofenac, metoprolol, bisphenol A, 17β-estradiol) in water samples by GC/MS with selected ion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service