Skip to Content
Merck
All Photos(1)

Key Documents

32343

Supelco

Caspofungin diacetate

analytical standard

Synonym(s):

1-[(4R,5S)-5-[(2-Aminoethyl)amino]-N2-(10,12-dimethyl-1-oxotetradecyl)-4-hydroxy-L-ornithine]-5-[(3R)-3-hydroxy-L-ornithine]-pneumocandin B0

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€98.90

€98.90


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€98.90

About This Item

Empirical Formula (Hill Notation):
C52H88N10O15 · 2C2H4O2
CAS Number:
Molecular Weight:
1213.42
UNSPSC Code:
41116107
NACRES:
NA.24

€98.90


Estimated to ship on25 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC([O-])=O.CC([O-])=O.CC[C@H](C)C[C@H](C)CCCCCCCCC(=O)N[C@H]1C[C@@H](O)[C@@H](NCC[NH3+])NC(=O)[C@@H]2[C@@H](O)CCN2C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H]3C[C@@H](O)CN3C(=O)[C@@H](NC1=O)[C@@H](C)O)[C@H](O)[C@@H](O)c4ccc(O)cc4)[C@H](O)CC[NH3+]

InChI

1S/C52H88N10O15.2C2H4O2/c1-5-28(2)24-29(3)12-10-8-6-7-9-11-13-39(69)56-34-26-38(68)46(55-22-21-54)60-50(75)43-37(67)19-23-61(43)52(77)41(36(66)18-20-53)58-49(74)42(45(71)44(70)31-14-16-32(64)17-15-31)59-48(73)35-25-33(65)27-62(35)51(76)40(30(4)63)57-47(34)72;2*1-2(3)4/h14-17,28-30,33-38,40-46,55,63-68,70-71H,5-13,18-27,53-54H2,1-4H3,(H,56,69)(H,57,72)(H,58,74)(H,59,73)(H,60,75);2*1H3,(H,3,4)/t28-,29+,30+,33+,34-,35-,36+,37-,38+,40-,41-,42-,43-,44-,45-,46-;;/m0../s1

InChI key

OGUJBRYAAJYXQP-IJFZAWIJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Caspofungin is an echinocandin antifungal that inhibits 1,3-β-D-glucan synthase which is required for cell wall synthesis.
Caspofungin is an echinocandin antifungal that inhibits 1,3-β-D-glucan synthase, which is required for cell wall synthesis, thereby disturbing the integrity of the fungal cell wall.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jitka Kočendová et al.
FEMS yeast research, 19(3) (2019-02-13)
Candida albicans is the main causative agent of vulvovaginal candidiasis (VVC), a common mycosis in women, relapses of which are difficult to manage due to biofilm formation. This study aimed at developing novel non-toxic compounds active against Candida spp. biofilms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service