Skip to Content
Merck
All Photos(3)

Key Documents

323101

Sigma-Aldrich

2-Methyl-2-adamantanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

powder or crystals

mp

215-217 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC1(O)[C@H]2C[C@@H]3C[C@H](C2)C[C@H]1C3

InChI

1S/C11H18O/c1-11(12)9-3-7-2-8(5-9)6-10(11)4-7/h7-10,12H,2-6H2,1H3/t7-,8+,9-,10+,11?

InChI key

JKOZWMQUOWYZAB-XICIVTQQSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanlei Xie et al.
Analytical and bioanalytical chemistry, 411(10), 2131-2140 (2019-02-06)
Herein, we developed a sensitive and quantitative flow assay for simultaneous detection of amantadine (AMD) and chloramphenicol (CAP) in chicken samples based on different CdSe/ZnS quantum dots (QDs). In contrast to other reports, the QDs could be excited by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service