Skip to Content
Merck
All Photos(2)

Key Documents

32233

Supelco

Danofloxacin-(methyl-d3)

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D3H17FN3O3
Molecular Weight:
360.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[2H]C([2H])([2H])N1C[C@@H]2C[C@H]1CN2c3cc4N(C=C(C(O)=O)C(=O)c4cc3F)C5CC5

General description

Chemical structure: quinolone

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thirayost Nimmanon et al.
Cellular and molecular life sciences : CMLS, 78(4), 1781-1798 (2020-08-17)
Zinc has been known to be essential for cell division for over 40 years but the molecular pathways involved remain elusive. Cellular zinc import across biological membranes necessitates the help of zinc transporters such as the SLC39A family of ZIP transporters.
Caiyong Ye et al.
Nucleic acids research, 48(22), 12817-12832 (2020-11-28)
Ribosome biogenesis is a fundamental process required for cell proliferation. Although evolutionally conserved, the mammalian ribosome assembly system is more complex than in yeasts. BCCIP was originally identified as a BRCA2 and p21 interacting protein. A partial loss of BCCIP
Xueqian Gong et al.
Cancer discovery, 9(2), 248-263 (2018-10-31)
Loss-of-function mutations in the retinoblastoma gene RB1 are common in several treatment-refractory cancers such as small-cell lung cancer and triple-negative breast cancer. To identify drugs synthetic lethal with RB1 mutation (RB1mut), we tested 36 cell-cycle inhibitors using a cancer cell
Changhan Ouyang et al.
Autophagy, 16(10), 1753-1770 (2019-12-28)
Macroautophagy/autophagy, a fundamental process for degradation of macromolecules and organelles, occurs constitutively at a basal level and is upregulated in response to stress. Whether autophagy regulates protein acetylation and microtubule stability in vascular smooth muscle cells (VSMCs) migration, however, remains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service