Skip to Content
Merck
All Photos(2)

Key Documents

32174

Supelco

Buquinolate

VETRANAL®, analytical standard

Synonym(s):

4-Hydroxy-6,7-diisobutoxyquinoline-3-carboxylic acid ethyl ester, Ethyl 4-hydroxy-6,7-diisobutoxyquinoline-3-carboxylate, NSC 201116

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€650.00

€650.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€650.00

About This Item

Empirical Formula (Hill Notation):
C20H27NO5
CAS Number:
Molecular Weight:
361.43
Beilstein:
493276
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€650.00


Estimated to ship on26 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

n1c2c(c(c(c1)C(=O)OCC)O)cc(c(c2)OCC(C)C)OCC(C)C

InChI

1S/C20H27NO5/c1-6-24-20(23)15-9-21-16-8-18(26-11-13(4)5)17(25-10-12(2)3)7-14(16)19(15)22/h7-9,12-13H,6,10-11H2,1-5H3,(H,21,22)

InChI key

LVVXOXRUTDAKFE-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takayuki Nakajima et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 52(3), 178-182 (2011-07-02)
We studied the simultaneous determination of nequinate and buquinolate, which are used as feed additives to prevent coccidiosis, by means of liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS). The sample was extracted with acetonitrile, then loaded onto an HLB

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service