Skip to Content
Merck
All Photos(1)

Key Documents

32131

Supelco

Fluacrypyrim

PESTANAL®, analytical standard

Synonym(s):

E)-α-(Methoxymethylene-2-[[[2-(1-methylethoxy)-6-(trifluoromethyl)-4-pyrimidinyl]oxy]methyl]-benzeneacetic acid methyl ester, Methyl (E)-2-{α-[2-isopropoxy-6-(trifluoromethyl)pyrimidin-4-yloxy]-o-tolyl}-3-methoxyacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21F3N2O5
CAS Number:
Molecular Weight:
426.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CO\C=C(\C(=O)OC)c1ccccc1COc2cc(nc(OC(C)C)n2)C(F)(F)F

InChI

1S/C20H21F3N2O5/c1-12(2)30-19-24-16(20(21,22)23)9-17(25-19)29-10-13-7-5-6-8-14(13)15(11-27-3)18(26)28-4/h5-9,11-12H,10H2,1-4H3/b15-11+

InChI key

MXWAGQASUDSFBG-RVDMUPIBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluacrypyrim is a novel small molecule, capable of inhibiting STAT3 protein activation in the G1 phase of cell cycle, thereby inducing cell cycle arrest, hence preventing growth of leukemia cells.[1] It has a very high selectivity for inhibiting aflatoxin produced by the aflatoxigenic fungus.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitory effects of respiration inhibitors on aflatoxin production.
Toxins, 6 (4), 1193-1200 (2014)
Fluacrypyrim, a novel STAT3 activation inhibitor, induces cell cycle arrest and apoptosis in cancer cells harboring constitutively-active STAT3.
International Journal of Cancer. Journal International Du Cancer, 127 (6), 1259-1270 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service