Skip to Content
Merck
All Photos(2)

Key Documents

32123

Supelco

Imidocarb dihydrochloride monohydrate

VETRANAL®, analytical standard

Synonym(s):

N,N′-Bis[3-(2-imidazolin-2-yl)phenyl]urea dihydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N6O · 2HCl · H2O
CAS Number:
Molecular Weight:
439.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O.Cl.Cl.O=C(Nc1cccc(c1)C2=NCCN2)Nc3cccc(c3)C4=NCCN4

InChI

1S/C19H20N6O.2ClH.H2O/c26-19(24-15-5-1-3-13(11-15)17-20-7-8-21-17)25-16-6-2-4-14(12-16)18-22-9-10-23-18;;;/h1-6,11-12H,7-10H2,(H,20,21)(H,22,23)(H2,24,25,26);2*1H;1H2

InChI key

ZZMCPNKUQHIZDQ-UHFFFAOYSA-N

General description

Imidocarb dihydrochloride monohydrate, (CAS:5318-76-3 (anhydrous)) VETRANAL analytical standard is a high purity veterinary drug standard used for the determination of the active ingredients of the drug in processed food of animal origin.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Barré et al.
Ticks and tick-borne diseases, 2(1), 55-61 (2011-07-21)
In December 2007, Babesia bovis was introduced to New Caledonia through the importation of cattle that had been vaccinated with a live tick fever (babesiosis and anaplasmosis) vaccine. Although the tick Rhipicephalus (Boophilus) microplus is common in New Caledonia, the
A F Lehner et al.
Journal of analytical toxicology, 35(4), 199-204 (2011-04-26)
Imidocarb [N,N'-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea, C(19)H(20)N(6)O(1), m.w. 348.41] is a symmetrical carbanilide derivative used to treat disease caused by protozoans of the Babesia genus. Imidocarb, however, is also considered capable of suppressing Babesia-specific immune responses, allowing Babesia-positive horses to pass a complement fixation
Øivind Øines et al.
Veterinary parasitology, 171(3-4), 350-353 (2010-04-10)
An Irish setter from the Oslo area was presented to the clinic with signs of babesiosis, a few days after a tick bite. Blood analysis confirmed babesiosis. Microscopic examination of thin blood film revealed large, basophilic, bodies inside erythrocytes, indicative
Huimin Yu et al.
Se pu = Chinese journal of chromatography, 29(8), 750-754 (2011-12-02)
A quantitive method for the determination of imidocarb residue in edible tissues of swine by high performance liquid chromatography (HPLC) was developed. The analyte was digested by beta-glucosidase first, and then extracted with 1 mol/L hydrochloric acid. The aqueous phase
Robin W Allison et al.
Veterinary clinical pathology, 40(3), 345-350 (2011-07-28)
A 5-month-old intact male Boerboel dog, imported from South Africa 1 week previously, was presented to a Texas veterinarian for lethargy, anorexia, and labored breathing. The dog was febrile, anemic, leukopenic, thrombocytopenic, and slightly azotemic. Results of the IDEXX SNAP-4Dx

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service