Skip to Content
Merck
All Photos(1)

Key Documents

32122

Supelco

5′-Hydroxypiroxicam

VETRANAL®, analytical standard

Synonym(s):

4-Hydroxy-N-(5-hydroxy-2-pyridyl)-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O5S
CAS Number:
Molecular Weight:
347.35
Beilstein:
5142716
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CN1C(C(=O)Nc2ccc(O)cn2)=C(O)c3ccccc3S1(=O)=O

InChI

1S/C15H13N3O5S/c1-18-13(15(21)17-12-7-6-9(19)8-16-12)14(20)10-4-2-3-5-11(10)24(18,22)23/h2-8,19-20H,1H3,(H,16,17,21)

InChI key

CCKOORANQAQKKV-UHFFFAOYSA-N

General description

5′-Hydroxypiroxicam (CAS:77459-78-0) belongs to the VETRANAL product line of high purity veterinary drug standards used for the determination of the active ingredients of the drug in processed food of animal origin.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Klopas et al.
Journal of pharmaceutical and biomedical analysis, 17(3), 515-524 (1998-07-10)
A zero-crossing first-derivative spectrophotometric method for the determination of piroxicam and its major metabolite 5-hydroxypiroxicam (5-HP) in human plasma is described. This technique permits the quantification of compounds with closely overlapping spectral bands without any separation step. The method consists
High-performance liquid chromatographic analysis of piroxicam and its major metabolite 5'-hydroxypiroxicam in human plasma and urine.
C J Richardson et al.
Journal of chromatography, 382, 382-388 (1986-10-31)
V Ródenas et al.
The Analyst, 123(8), 1749-1752 (1999-03-11)
A first-derivative spectrophotometric method for the simultaneous determination of piroxicam (PX) and its major metabolite 5'-hydroxypiroxicam (OH PX) in human plasma is described. The method consists of direct extraction of the two drugs from the plasma samples with hydrochloric and
C J Richardson et al.
European journal of clinical pharmacology, 32(1), 89-91 (1987-01-01)
Piroxicam (20 mg once daily) was administered orally to six healthy young volunteers for 15 days. Trough steady-state levels of piroxicam and 5'-hydroxypiroxicam were 5.5 and 1.2 micrograms/ml, respectively. Piroxicam's plasma half-life (54.9 h) was significantly shorter than that of
P A Milligan
Journal of chromatography, 576(1), 121-128 (1992-04-15)
A simple and sensitive liquid chromatographic method with ultraviolet detection is described for the determination of the nonsteroidal anti-inflammatory drug piroxicam and its major metabolites in human plasma, urine and bile. Separation of these components occurs on a reversed phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service