Skip to Content
Merck
All Photos(1)

Key Documents

31735

Supelco

Benzathine penicilline G tetrahydrate

VETRANAL®, analytical standard

Synonym(s):

Benzylpenicillin Benzathin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H56N6O8S2 · 4H2O
CAS Number:
Molecular Weight:
981.18
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

O.O.O.O.C(CNCc1ccccc1)NCc2ccccc2.[H][C@]34SC(C)(C)[C@@H](N3C(=O)[C@H]4NC(=O)Cc5ccccc5)C(O)=O.[H][C@]67SC(C)(C)[C@@H](N6C(=O)[C@H]7NC(=O)Cc8ccccc8)C(O)=O

InChI

1S/2C16H18N2O4S.C16H20N2.4H2O/c2*1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16;;;;/h2*3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);1-10,17-18H,11-14H2;4*1H2/t2*11-,12+,14-;;;;;/m11...../s1

InChI key

WIDKTXGNSOORHA-CJHXQPGBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn Margaret Dupnik et al.
Leprosy review, 83(4), 389-393 (2013-04-26)
There are diverse skin manifestations of both leprosy and syphilis. These diseases can appear similar to many other dermatologic conditions as well as to systemic diseases with dermatologic signs. Nodular syphilis is an uncommon type of secondary syphilis. We present
Syphilis with a lichen planus-like pattern (hypertrophic syphilis).
Gianluca Nazzaro et al.
Journal of cutaneous pathology, 39(8), 805-807 (2012-07-04)
Piasecka Katarzyna et al.
Klinika oczna, 114(1), 50-52 (2012-07-13)
Syphilis is a sexually transmitted, chronic, systemic infection of increasing prevalence. Ocular symptoms may develop at any stage of the disease process. This is to present a case of acute severe bilateral visual loss due to neuroretinitis with papillar oedema
A crusted plaque on the right nipple.
Hsien-Yi Chiu et al.
JAMA, 308(4), 403-404 (2012-07-24)
An uncommon cause of ulceroinfiltrative gastric wall thickening in a young patient.
Young Chul Kim et al.
Gastroenterology, 143(5), e6-e7 (2012-09-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service