Skip to Content
Merck
All Photos(1)

Key Documents

31715

Supelco

Hexachloroethane

VETRANAL®, analytical standard

Synonym(s):

Perchloroethane

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
Cl3CCCl3
CAS Number:
Molecular Weight:
236.74
Beilstein:
1740341
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

8.16 (vs air)

vapor pressure

0.4 mmHg ( 20 °C)

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

183-185 °C (dec.) (lit.)

density

2.091 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC(Cl)(Cl)C(Cl)(Cl)Cl

InChI

1S/C2Cl6/c3-1(4,5)2(6,7)8

InChI key

VHHHONWQHHHLTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexachloroethane may be used as a reference standard for the determination of the analyte in environmental samples by high-performance liquid chromatography (HPLC)[1] and selected-ion monitoring gas chromatography-mass spectrometry.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative analysis of polychlorinated biphenyls, organochlorine insecticides, polycyclic aromatic hydrocarbons, polychlorinated hydrocarbons and polynitrohydrocarbons in spiked samples of soil, water and plasma by selected-ion monitoring gas chromatography- mass spectrometry.
Singh AK, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 706(2), 231- 244 (1998)
Quantitative determination of several simple perhalogenated compounds by high-performance liquid chromatography.
Collins CH and Morgano MA
Journal of Chromatography A, 846(1-2), 395- 399 (1999)
E V Patterson et al.
Journal of the American Chemical Society, 123(9), 2025-2031 (2001-07-18)
Ab initio and density functional levels of electronic structure theory are applied to characterize alternative mechanisms for the reductive dechlorination of hexachloroethane (HCA) to perchloroethylene (PCE). Aqueous solvation effects are included using the SM5.42R continuum solvation model. After correction for
Ching-Hui Loh et al.
Toxicology, 247(2-3), 119-122 (2008-04-18)
There has been no human epidemiological data regarding the hepatic injuries of hexachloroethane-zinc oxide (HC/ZnO) inhalation. This is the first epidemiological study to investigate whether HC/ZnO inhalation exposure can induce hepatic dysfunction in exposed soldiers. Twenty soldiers, exposed to high
P S Holmes
Military medicine, 164(10), 751-752 (1999-11-02)
Hexachloroethane (HC) smoke, also known as white smoke, is an obscurant used in numerous military situations. Many adverse health effects are associated with the use of white smoke, some of which are potentially life threatening. Inhalation is the most frequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service