Skip to Content
Merck
All Photos(1)

Documents

31534

Supelco

2-Imidazolidone

PESTANAL®, analytical standard

Synonym(s):

N,N′-Ethyleneurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2O
CAS Number:
Molecular Weight:
86.09
Beilstein:
106252
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

129-132 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

O=C1NCCN1

InChI

1S/C3H6N2O/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)

InChI key

YAMHXTCMCPHKLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Find more information here: Neonicotinoids

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Residues of mancozeb, 2-imidazoline, and ethyleneurea in tomato and potato crops after field treatment with mancozeb.
W H Newsome
Journal of agricultural and food chemistry, 27(6), 1188-1190 (1979-11-01)
D A Kuznetsov et al.
Cancer letters, 34(1), 61-66 (1987-01-01)
Daily intragastric administration of a carcinogen N,N'-ethylenethiourea (ETU) (85 mg/kg body wt, congruent to 0.1 DL50) leads to significant polyadenylate polymerase (PAP) activity in rat blood serum by the 10th day of experiment. A similar course of N,N'-ethyleneurea (EU) fails
[Polyadenylate polymerase in the diagnosis of the early stages of chemical carcinogenesis].
N V Zav'ialov et al.
Gigiena truda i professional'nye zabolevaniia, (2)(2), 25-28 (1988-02-01)
Shelley L Aldrich et al.
Military medicine, 176(5), 584-585 (2011-06-04)
Ethylene urea/melamine formaldehyde resin (permanent press) is a common fabric finishing agent added to Army Combat Uniforms for a wrinkle-free appearance and to strengthen the fabric. We describe the case of an active duty U.S. Army soldier with a diffuse
Julie A Spicer et al.
Journal of medicinal chemistry, 56(23), 9542-9555 (2013-11-08)
A series of novel 5-arylidene-2-thioxoimidazolidin-4-ones were investigated as inhibitors of the lymphocyte-expressed pore-forming protein perforin. Structure-activity relationships were explored through variation of an isoindolinone or 3,4-dihydroisoquinolinone subunit on a fixed 2-thioxoimidazolidin-4-one/thiophene core. The ability of the resulting compounds to inhibit

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service