Skip to Content
Merck
All Photos(1)

Key Documents

31405

Millipore

Dextrin from potato starch

for biotechnological purposes

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€61.40

€61.40


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€61.40

About This Item

Linear Formula:
(C6H12O6)x
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

€61.40


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

form

powder

solubility

H2O: 0.1 g/mL, clear, yellow to very deep yellow (hot)

application(s)

microbiology

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Toshikazu Sugimoto et al.
Journal of industrial microbiology & biotechnology, 38(12), 1985-1991 (2011-05-28)
Submerged batch cultures of Aspergillus kawachii grown on indigestible dextrin were investigated for potential improvements in glucoamylase (GA) production. In flask culture, specific GA productivities per dry weight biomass using dextrin and indigestible dextrin were 11.0 and 56.1 mU/mg-DW, respectively. Indigestible
Glucose polymer supplements in low birth weight infants.
O G Brooke
British medical journal (Clinical research ed.), 286(6371), 1143-1143 (1983-04-02)
Galina A Kovalenko et al.
Carbohydrate research, 343(7), 1202-1211 (2008-03-19)
Glucoamylase (GA) was immobilized by adsorption on carbon support: on Sibunit, on bulk catalytic filamentous carbon (bulk CFC) and on activated carbon (AC). This was used to prepare heterogeneous biocatalysts for the hydrolysis of starch dextrin. The effect of the
Glucose-polymer in acute diarrhoea.
Archives of disease in childhood, 57(9), 721-722 (1982-09-01)
O E Odeleye et al.
European journal of clinical nutrition, 46(5), 367-371 (1992-05-01)
The effect of caloreen (glucose polymer) supplementation on indicators of iron status during protein-energy malnutrition was studied. Sixty-four children with moderate protein energy malnutrition (PEM) were fed diets supplemented with caloreen or starch (control) for 14 days, following which iron

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service