Skip to Content
Merck

Skip To

31385

Devarda′s alloy

puriss. p.a.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
AlCuZn
CAS Number:
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
41116105
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

puriss. p.a.

Quality Segment

form

powder

composition

aluminum, ~45% , copper, ~50% , zinc, ~5%

mp

550 °C (lit.)

application(s)

food and beverages
forensics and toxicology

SMILES string

[Al].[Cu].[Zn]

InChI

1S/Al.Cu.Zn

InChI key

MUBKMWFYVHYZAI-UHFFFAOYSA-N

General description

Devarda′s alloy is a reducing agent, and has found application in being used to reduce NO3--N, or NO2--N, into NH4+-N, thereby finding functionality in being used for total nitrogen determination.

Application

Reducing agent used for the determination of total nitrogen by the method of Kjeldahl. Reagent used for the reduction of alkaline solutions of chlorate to chloride
Devarda′s alloy was used to study, its effects as a reducing agent on Cp2TiCl-catalyzed living radical polymerization of styrene, which was initiated by epoxide radical ring opening.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4839072694841.05341
form

powder

form

powder

form

filings

form

powder

mp

550 °C (lit.)

mp

550 °C (lit.)

mp

550 °C (lit.)

mp

490-560 °C

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

300

application(s)

food and beverages
forensics and toxicology

application(s)

-

application(s)

-

application(s)

-

composition

aluminum, ~45% , zinc, ~5% , copper, ~50%

composition

aluminum, ~45% , copper, ~50% , zinc, ~5%

composition

aluminum, ~45% , copper, ~50% , zinc, ~5%

composition

-

grade

puriss. p.a.

grade

-

grade

-

grade

-


Still not finding the right product?

Explore all of our products under Devarda′s alloy


pictograms

FlameEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 2 - Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters