Skip to Content
Merck
All Photos(1)

Key Documents

31297

Supelco

PCB No 52 solution

10 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,5,5′-Tetrachlorobiphenyl solution, 2,2′,5,5′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl4
CAS Number:
Molecular Weight:
291.99
Beilstein:
2053828
Ballschmiter Number:
52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(Cl)c(c1)-c2cc(Cl)ccc2Cl

InChI

1S/C12H6Cl4/c13-7-1-3-11(15)9(5-7)10-6-8(14)2-4-12(10)16/h1-6H

InChI key

HCWZEPKLWVAEOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suleyman Sandal et al.
Immunopharmacology and immunotoxicology, 27(4), 601-613 (2006-01-27)
Polychlorinated biphenyls (PCBs) are industrial chemicals that have been released into the environment, resulting in widespread and persistent contamination. PCBs exist as 209 different congeners depending on the chlorine substitution on the biphenyl rings, and the physical properties and toxic
Chia-Hua Lin et al.
Toxicology letters, 188(1), 11-19 (2009-05-13)
Polychlorinated biphenyls (PCBs) are ubiquitous environmental contaminants. Much of the research has focused on the carcinogenic potential of higher chlorinated PCBs, but accumulative evidence has shown that lower chlorinated PCB congeners have initiating and promoting activities. The goal of this
Xinzhu Pu et al.
Toxicology, 217(1), 14-21 (2005-09-21)
The bioavailability of coplanar 2,3',4,4',5-pentachlorobiphenyl (PCB118) and nonplanar 2,2',5,5'-tetrachlorobiphenyl (PCB52) from soils representing a range in organic carbon (OC), clay content and pH were investigated using an in vivo rat model and an in vitro physiologically based extraction test (PBET)
Kristine L Richardson et al.
Chemical research in toxicology, 24(5), 718-725 (2011-04-13)
The rates of oxidative metabolism of two tetrachlorobiphenyl congeners were determined in hepatic microsomes from four species of sea turtles, green (Chelonia mydas), olive ridley (Lepidochelys olivacea), loggerhead (Caretta caretta), and hawksbill (Eretmochelys imbricata). Hydroxylation of 3,3',4,4'-tetrachlorobiphenyl (PCB 77), an
Lilin Wang et al.
Environmental science & technology, 46(10), 5422-5429 (2012-04-17)
Stable colloidal suspensions of buckminsterfullerene (nC(60)) in aqueous environments can significantly affect the fate and transport of hydrophobic organic contaminants by serving as a contaminant carrier. In this study, we examined enhanced transport of 2,2',5,5'-polychlorinated biphenyl (PCB) in saturated sandy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service