Skip to Content
Merck
All Photos(1)

Key Documents

30929

Supelco

Saxitoxin-15N7 dihydrochloride solution

10 μg/g in hydrochloric acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
€325.00

€325.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
0.5 ML
€325.00

About This Item

Empirical Formula (Hill Notation):
C10H1715N7O4 · 2HCl
Molecular Weight:
379.16
UNSPSC Code:
41116107
NACRES:
NA.24

€325.00


Estimated to ship on15 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

10 μg/g in hydrochloric acid (nominal concentration)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]3([15NH]C([15NH]2)=[15NH])[15N](CCC3(O)O)C([15NH][C@H]1COC([15NH2])=O)=[15NH]

InChI

1S/C10H17N7O4/c11-6-15-5-4(3-21-8(13)18)14-7(12)17-2-1-9(19,20)10(5,17)16-6/h4-5,19-20H,1-3H2,(H2,12,14)(H2,13,18)(H3,11,15,16)/t4-,5-,10-/m0/s1/i11+1,12+1,13+1,14+1,15+1,16+1,17+1

InChI key

RPQXVSUAYFXFJA-FXWZNOLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Paralytic Shellfish Toxin PST for the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materialsThis is the 13C isotope-labelled analog of saxitoxin.

Analysis Note

10 μg/g in 3 mmol/l HCl in water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service