Skip to Content
Merck
All Photos(1)

Key Documents

30888

Sigma-Aldrich

Gum arabic from acacia tree

spray dried, tested according to Ph. Eur.

Synonym(s):

Acacia gum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

biological source

plant (acacia)

Agency

tested according to Ph. Eur.

form

powder

solubility

water: soluble

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Acacia gum has been used as a surfactant in the development of hydrogels for tissue engineering applications. Acacia gum polymer has also been used to preserve microorganisms during sampling and storage.

Biochem/physiol Actions

Gum arabic (GA) is reported to be a strong anti-oxidant and has shown protection against nephrotoxicity in mice by the generation of free radicals. GA also reduced the blood glucose concentration by initiating the release of insulin from the pancreatic β cells. It has hypolipidemic effect by increasing fecal bile acid and modification of lipid digestion. It is a potential therapeutic agent in hepatic and renal failures. It contains enzymes like oxidases, pectinases and peroxidases that act against microbes that cause tooth decay. Gum arabic is implicated in adverse effects like suppression of macrophage activation.

Other Notes

Believed to be a branched polymer of galactose, rhamnose, arabinose, and glucuronic acid as the calcium, magnesium, and potassium salts with a mol. wt. of approx. 250,000.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B H Ali
Renal failure, 26(1), 1-3 (2004-04-16)
Gum Arabic has recently been claimed to be effective in preventing gentamicin-induced acute renal failure in rats. The nephroprotection was suggested to be due to an anti-oxidant action by the gum Arabic. In the present work we tested whether treatment
Badreldin H Ali et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 47(1), 1-8 (2008-08-02)
Gum arabic (GA) is a branched-chain, complex polysaccharide, either neutral or slightly acidic, found as a mixed calcium, magnesium and potassium salt of a polysaccharidic acid. The backbone is composed of 1,3-linked beta-D-galactopyranosyl units. The side chains are composed of
Iryna Sorokulova et al.
Journal of microbiological methods, 88(1), 140-146 (2011-11-19)
Stability of microbial cultures during sampling and storage is a vital issue in various fields of medicine, biotechnology, food science, and forensics. We have developed a unique bacterial preservation process involving a non-toxic, water-soluble acacia gum polymer that eliminates the
K Fujiwara et al.
Journal of ethnopharmacology, 46(2), 107-114 (1995-05-01)
The effect of Xiao-Chai-Hu-Tang (Sho-saiko-to, TJ-9), the extract of a mixture of 7 herbs, on hepatic macrophage function was studied using rats. Hepatic macrophages were activated by injection of Corynebacterium parvum or 70% partial hepatectomy. Oral administration of TJ-9 for
Chengdong Ji et al.
Biomaterials, 32(36), 9719-9729 (2011-09-20)
The aim of this study was to develop a process to create highly porous three-dimensional (3D) chitosan hydrogels suitable for tissue engineering applications. Chitosan was crosslinked by glutaraldehyde (0.5 vol %) under high pressure CO(2) at 60 bar and 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service