Skip to Content
Merck
All Photos(1)

Key Documents

30723

Sigma-Aldrich

Hydrochloric acid solution

puriss. p.a., ≥25%

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor pressure

190 mmHg ( 25 °C)

grade

puriss. p.a.

Assay

≥25%

impurities

residual solvents, complies
≤0.00005% free chlorine (Cl)
≤0.0001% ammonium (NH4)

ign. residue

≤0.0005% (as SO4)

bp

100 °C (lit.)

anion traces

bromide (Br-): ≤50 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤1 mg/kg
sulfite (SO32-): ≤1 mg/kg

cation traces

Al: ≤0.05 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.02 mg/kg
Be: ≤0.02 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.2 mg/kg
Ge: ≤0.05 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.02 mg/kg
Sr: ≤0.01 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrochloric acid solution, puriss. p.a., ≥25% is used as a high purity (>98.5%) analytical reagent.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE HYDROLYSIS OF CHITIN BY CONCENTRATED HYDROCHLORIC ACID, AND THE PREPARATION OF LOW-MOLECULAR-WEIGHT SUBSTRATES FOR LYSOZYME.
J A RUPLEY
Biochimica et biophysica acta, 83, 245-255 (1964-11-01)
Daejin Kim et al.
Environmental science & technology, 49(16), 9452-9459 (2015-06-25)
The rare earth elements (REEs) such as neodymium, praseodymium, and dysprosium were successfully recovered from commercial NdFeB magnets and industrial scrap magnets via membrane assisted solvent extraction (MSX). A hollow fiber membrane system was evaluated to extract REEs in a
Quantitative estimation of sialic acids. II. A colorimetric resorcinol-hydrochloric acid method.
L SVENNERHOLM
Biochimica et biophysica acta, 24(3), 604-611 (1957-06-01)
Brian W Simons et al.
The Journal of pathology, 235(3), 478-489 (2014-10-29)
Inflammation is associated with several diseases of the prostate including benign enlargement and cancer, but a causal relationship has not been established. Our objective was to characterize the prostate inflammatory microenvironment after infection with a human prostate-derived bacterial strain and
Jeffery A Goldstein et al.
Human molecular genetics, 23(25), 6722-6731 (2014-07-30)
Disruption of the dystrophin complex causes muscle injury, dysfunction, cell death and fibrosis. Excess transforming growth factor (TGF) β signaling has been described in human muscular dystrophy and animal models, where it is thought to relate to the progressive fibrosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service