Skip to Content
Merck
All Photos(1)

Key Documents

30160

Sigma-Aldrich

L-Cysteine methyl ester hydrochloride

puriss., ≥99.0% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
171.65
Beilstein:
3685824
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

puriss.

Assay

≥99.0% (RT)

optical activity

[α]20/D −2.1±0.2°, c = 10% in methanol

mp

~140 °C
142 °C (dec.) (lit.)

SMILES string

Cl[H].COC(=O)[C@@H](N)CS

InChI

1S/C4H9NO2S.ClH/c1-7-4(6)3(5)2-8;/h3,8H,2,5H2,1H3;1H/t3-;/m0./s1

InChI key

WHOHXJZQBJXAKL-DFWYDOINSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Used e.g. in the synthesis of peptides; Preparation of thiazolidine-4-carboxylates with carbonyl compounds

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.E. Baldwin et al.
Tetrahedron Letters, 27, 3461-3461 (1986)
D. Chiarino et al.
Journal of Heterocyclic Chemistry, 26, 589-589 (1989)
Y. Hamada et al.
The Journal of Organic Chemistry, 52, 1252-1252 (1987)
M.D. Threadgill et al.
The Journal of Organic Chemistry, 54, 2940-2940 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service