Skip to Content
Merck
All Photos(1)

Key Documents

29598

Supelco

Cyclooctane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16
CAS Number:
Molecular Weight:
112.21
Beilstein:
1900349
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor pressure

16 mmHg ( 37.7 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.458 (lit.)
n20/D 1.458

bp

151 °C/740 mmHg (lit.)

mp

10-13 °C (lit.)

density

0.834 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

C1CCCCCCC1

InChI

1S/C8H16/c1-2-4-6-8-7-5-3-1/h1-8H2

InChI key

WJTCGQSWYFHTAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pieter van Delft et al.
Organic letters, 12(23), 5486-5489 (2010-11-06)
The conjugation of a ribonucleic acid 16-mer with the cationic amphiphilic peptide penetratin and an anionic hyaluronan tetrasaccharide by means of Cu-free "click" chemistry is reported. The alkyne-functionalized 16-mer was prepared by automated solid-phase synthesis, using a newly developed strained
Shawn Martin et al.
The Journal of chemical physics, 132(23), 234115-234115 (2010-06-25)
Understanding energy landscapes is a major challenge in chemistry and biology. Although a wide variety of methods have been invented and applied to this problem, very little is understood about the actual mathematical structures underlying such landscapes. Perhaps the most
Heidi E Bostic et al.
Chemical communications (Cambridge, England), 48(10), 1431-1433 (2011-10-04)
Copper-free click chemistry was employed to derivatize membrane bilayers. This approach uses an azido-lipid conjugate presented on liposomes, which can be labeled in bioorthogonal fashion via cyclooctyne-tagged reagents. An immobilization-based approach using streptavidin-coated microplates was exploited to evaluate membrane derivatization.
Leo A Paquette et al.
The Journal of organic chemistry, 71(12), 4353-4363 (2006-06-06)
D-Arabinose and D-glucose are transformed into the identical vinyl furanoside, whose role is to serve as the precursor to enantiopure cyclooctadienone 6. The key steps of this relay involve a zirconocene-promoted ring contraction and [3,3] sigmatropic rearrangement of an enynol.
W Michael Brown et al.
The Journal of chemical physics, 129(6), 064118-064118 (2008-08-22)
Dimensionality reduction approaches have been used to exploit the redundancy in a Cartesian coordinate representation of molecular motion by producing low-dimensional representations of molecular motion. This has been used to help visualize complex energy landscapes, to extend the time scales

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service