Skip to Content
Merck
All Photos(2)

Documents

29242

Supelco

(−)-Guaiol

analytical standard

Synonym(s):

(3R,6S,10S)-6,10,α,α-Tetramethylbicyclo[5.3.0]dec-1(7)-ene-3-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein:
2047993
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

optical activity

[α]20/D −30.0±2°, c = 1 in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

91-93 °C (lit.)
91-93 °C

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1CC[C@H](CC2=C1CC[C@@H]2C)C(C)(C)O

InChI

1S/C15H26O/c1-10-5-7-12(15(3,4)16)9-14-11(2)6-8-13(10)14/h10-12,16H,5-9H2,1-4H3/t10-,11-,12+/m0/s1

InChI key

TWVJWDMOZJXUID-SDDRHHMPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-Guaiol is a sesquiterpene alcohol which has guaiane skeleton. It naturally occurs in essential oils extracted from Salvia lanigera and Helitta ongifoliata and has antibacterial activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microbial transformation of (-)-guaiol and antibacterial activity of its transformed products.
Choudhary MI
Journal of Natural Products, 70(5), 849-852 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service