Skip to Content
Merck
All Photos(1)

Key Documents

26411

Sigma-Aldrich

Calcium stearate

6.6-7.4% Ca basis

Synonym(s):

Octadecanoic acid calcium salt, Stearic acid calcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€57.40
20 KG
Inquire

€57.40


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€57.40
20 KG
Inquire

About This Item

Linear Formula:
[CH3(CH2)16COO]2Ca
CAS Number:
Molecular Weight:
607.02
Beilstein:
3919698
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes

€57.40


Estimated to ship on26 May 2025


Request a Bulk Order

grade

for analytical purposes

Quality Level

concentration

6.6-7.4% Ca

impurities

≤0.004% heavy metals (as Pb)
≤0.3% free acid (as stearic acid)

ign. residue

≤10%

loss

≤3% loss on drying, 105 °C, 3 h

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤1000 mg/kg

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Ca]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Ca/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

CJZGTCYPCWQAJB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphic transformations of calcium stearate and calcium stearate monohydrate.
R D VOLD et al.
Journal of colloid science, 3(4), 339-361 (1948-08-01)
Lorenzo Vannozzi et al.
Journal of the mechanical behavior of biomedical materials, 75, 147-159 (2017-07-25)
The fabrication of biomaterials for interaction with muscle cells has attracted significant interest in the last decades. However, 3D porous scaffolds featured by a relatively low stiffness (almost matching the natural muscle one) and highly stable in response to cyclic
Stearic acid and calcium stearate monolayer collapse.
Neuman RD.
Journal of Colloid and Interface Science, 56(3), 505-510 (1976)
Molecular orientation in the physical-vapour-deposited thin films of calcium stearate.
Miki T, et al.
Jpn J. App. Phys., Part I, 24(8A), L672-L672 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service