Skip to Content
Merck
All Photos(1)

Documents

26375

Sigma-Aldrich

4-Chloro-7-sulfobenzofurazan ammonium salt

for fluorescence, ≥99.0% (TLC)

Synonym(s):

4-Chloro-7-sulfobenz-2-oxa-1,3-diazole ammonium salt, 7-Chloro-2,1,3-benzoxadiazole-4-sulfonic acid ammonium salt, 7-Chloro-4-benzofurazansulfonic acid ammonium salt, SBF-Chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3ClN2O4S · NH3
CAS Number:
Molecular Weight:
251.65
Beilstein:
5200152
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for fluorescence

Assay

≥99.0% (TLC)

mp

>300 °C (lit.)

fluorescence

λex 383 nm; λem 525 nm in 0.1 M borate pH 9.0 (after derivatization with glutathione)

SMILES string

N.OS(=O)(=O)c1ccc(Cl)c2nonc12

InChI

1S/C6H3ClN2O4S.H3N/c7-3-1-2-4(14(10,11)12)6-5(3)8-13-9-6;/h1-2H,(H,10,11,12);1H3

InChI key

QFHXPBLOMHGYOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For the specific fluorescent determination of thiols

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium 4-chloro-7-sulfobenzofurazan: a new fluorigenic thiol-specific reagent.
J L Andrews et al.
Archives of biochemistry and biophysics, 214(1), 386-396 (1982-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service