Skip to Content
Merck
All Photos(1)

Key Documents

257397

Sigma-Aldrich

1,4-Bis(5-phenyl-2-oxazolyl)benzene

≥99%

Synonym(s):

1,4-Bis(5-phenyl-2-oxazolyl)benzene, 2,2′-p-Phenylene-bis(5-phenyloxazole), Di(phenyl-5-oxazolyl-2)-1,4-benzene, POPOP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N2O2
CAS Number:
Molecular Weight:
364.40
Beilstein:
325741
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥99%

mp

242-246 °C (lit.)

λmax

362 nm

SMILES string

c1ccc(cc1)-c2cnc(o2)-c3ccc(cc3)-c4ncc(o4)-c5ccccc5

InChI

1S/C24H16N2O2/c1-3-7-17(8-4-1)21-15-25-23(27-21)19-11-13-20(14-12-19)24-26-16-22(28-24)18-9-5-2-6-10-18/h1-16H

InChI key

MASVCBBIUQRUKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable as laser dye

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service