Skip to Content
Merck
All Photos(1)

Key Documents

256501

Sigma-Aldrich

Calcium carbonate

ACS reagent, ≥99.0%, low in alkalies

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent

Assay

≥99.0%

impurities

≤0.01% NH4OH pptd.
≤0.01% insol. dil. HCl

density

2.93 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
fluoride (F-): ≤0.0015%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.01%
Fe: ≤0.002%
K: ≤0.01%
Mg: ≤0.01%
NH4+: ≤0.003%
Na: ≤0.01%
Sr: ≤0.1%
heavy metals (as Pb): ≤0.001%

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From quarry to temple.
Heather Pringle
Science (New York, N.Y.), 340(6134), 805-805 (2013-05-21)
Guoqing Zhang et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 2046-2049 (2013-03-19)
The polymorphic phase transformation of thermally treated pearl powder was investigated by X-ray diffraction and thermoanalytical techniques. The phase transformation was based on quantification of the calcite content at various temperatures using Rietveld refinement analysis. The results show that the
Johannes Ihli et al.
Chemical communications (Cambridge, England), 49(30), 3134-3136 (2013-03-13)
A simple synthetic method is presented for the precipitation of high purity, dry amorphous calcium carbonate (ACC) based on freeze-drying saturated, counter ion free CaCO3 solutions, where the ACC produced shows an extended atmospheric stability. Translation of the methodology to
Yufei Ma et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 2413-2420 (2013-03-19)
We report an interesting finding of calcium carbonate (CaCO3) crystal growth in the silk fibroin (SF) hydrogel with different concentrations by a simple ion diffusion method. The experimental results indicate that the CaCO3 crystals obtained from silk fibroin gels with
Response to Annweiler and Beauchet.
Rebecca C Rossom et al.
Journal of the American Geriatrics Society, 61(6), 1050-1051 (2013-06-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service